Order Entry
Germany
ContactUsLinkComponent
Gel filtration chromatography columns, HiLoad™ with Superdex™ Prep Grade
Gel filtration chromatography columns, HiLoad™ with Superdex™ Prep Grade
Supplier:  Cytiva
Certificates
undefined
Gel filtration chromatography columns, HiLoad™ with Superdex™ Prep Grade
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 90-1001-37
  • 90-1002-73
  • 90-1000-40
  • Max. pressure:
  • 3 bar
  • 3 bar
  • 3 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Related Information