Protease inhibitor mix
Catalog # 80-6501-23
Supplier: Cytiva
CAS Number:
Specifications
- Pk:1 KIT
- Description:Protease inhibitor mix
Specifications
About this item
Sample preparation often requires the inhibition of protease activity. Protease inhibitor mix combines competitive and non competitive protease inhibitors, which protect proteins from proteolysis during purification from animal tissues, plant tissues, yeast and bacteria.
- Protease inhibitor cocktail specifically developed for sample preparation in 2-D studies
- No EDTA is used, which allows optimal nuclease activity for removing nucleic acids from samples
- Contains a mixture of both competitive and noncompetitive protease inhibitors that inhibit serine, cysteine, and calpain proteases - optionally, EDTA may be added to inhibit metalloproteases
- Optimised concentration for excellent inhibition of protease activities
- Effectively inhibits over 95% of protease activity
Protease inhibitor mix combines competitive and non competitive protease inhibitors, which protect proteins from proteolysis during purification from animal tissues, plant tissues, yeast and bacteria.