Order Entry
Germany
ContactUsLinkComponent
Gel filtration columns, HiPrep™ with Sephacryl® S-500 HR
Gel filtration columns, HiPrep™ with Sephacryl® S-500 HR
Supplier:  Cytiva
Certificates
undefined
Gel filtration columns, HiPrep™ with Sephacryl® S-500 HR
Supplier:  Cytiva

Specifications

  • Catalog No:
  • 28-9356-07
  • 28-9356-06
  • Max. pressure:
  • 1,5 bar
  • 1,5 bar
  • pH range:
  • 3 - 11
  • 3 - 11
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications