Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
Watch Glasses
Watch dishes
Watch dishes
:
VITLAB
Watch dishes
:
VITLAB
Watch dishes, made of PTFE, white.
Without base
Very good chemical resistance
Good temperature resistance
Ideal for use to cover beakers.
:
DIN EN 285