Order Entry
Germany
Orders LinkContactUsLinkComponent
Affinity chromatography columns, GSTrap™ FF

Affinity chromatography columns, GSTrap™ FF

Supplier:   Cytiva
Affinity chromatography columns, GSTrap™ FF
Supplier:  Cytiva
Certificates
Affinity chromatography columns, GSTrap™ FF

4 Options Available Below

Product Details & Documents

GSTrap FF columns are prepacked Glutathione Sepharose Fast Flow columns for fast, convenient, one-step purification of glutathione S-transferase (GST) tagged proteins.

  • Fast and simple one-step purification of GST-tagged proteins
  • Mild elution conditions preserving protein antigenicity and function
  • Easy scale-up by connecting the columns in series
  • Sepharose 4 Fast Flow provides good flow properties

GST-tagged proteins can be purified directly from pretreated bacterial lysates using GSTrap FF.GST-tagged proteins are eluted under mild, nondenaturing conditions using reduced glutathione.

The purification process preserves protein antigenicity and function. If desired, cleavage of the protein from GST can be achieved using a site-specific protease whose recognition sequence is located immediately upstream from the multiple cloning site on the pGEX plasmids. GST-tagged proteins can be detected using colorimetric or immunological methods.

The resin, Glutathione Sepharose 4 Fast Flow, is also available as lab packages and is an excellent choice for scale-up.The columns can be operated with a syringe, peristaltic pump, or liquid chromatography system such as ÄKTA design or FPLC System.

Specifications for Product Family

Specifications

  • Catalog No:
  • 17-5130-02
  • 17-5130-01
  • 17-5131-01
  • 17-5131-02
  • Max. pressure:
  • 5 bar
  • 5 bar
  • 5 bar
  • 5 bar
  • pH range:
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • 3 - 12
  • Storage:
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol
  • 4 to 30 °C, 20% Ethanol

Specifications

Product Family Options

Product Information

  • Column hardware
    Particle Size
    Bed dimensions
    Capacity
    Flow rate
    Flow velocity
    Availability
    Price
  • 17-5130-02
    GSTrap™ FF
    GSTrap™ FF
    PP
    90 µm
    7×25 mm
    2×1 ml
    <4 ml/min
    <450 cm/h
    Log in for availability
    Log in for availability
    1 * 2 ST
    1 * 2 ST - 187,00 €
    187,00 €
    1 * 2 ST
       187,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    17-5130-02
    Max. pressure
    5 bar
    pH range
    3 - 12
    Storage
    4 to 30 °C, 20% Ethanol
  • 17-5130-01
    GSTrap™ FF
    GSTrap™ FF
    PP
    90 µm
    7×25 mm
    5×1 ml
    <4 ml/min
    <450 cm/h
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 383,00 €
    383,00 €
    1 * 5 ST
       383,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    17-5130-01
    Max. pressure
    5 bar
    pH range
    3 - 12
    Storage
    4 to 30 °C, 20% Ethanol
  • 17-5131-01
    GSTrap™ FF
    GSTrap™ FF
    PP
    90 µm
    16×25 mm
    1×5 ml
    <20 ml/min
    <450 cm/h
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 365,00 €
    365,00 €
    1 * 1 ST
       365,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    17-5131-01
    Max. pressure
    5 bar
    pH range
    3 - 12
    Storage
    4 to 30 °C, 20% Ethanol
  • 17-5131-02
    GSTrap™ FF
    GSTrap™ FF
    PP
    90 µm
    16×25 mm
    5×5 ml
    <20 ml/min
    <450 cm/h
    Log in for availability
    Log in for availability
    1 * 5 ST
    1 * 5 ST - 1 450,00 €
    1 450,00 €
    1 * 5 ST
       1 450,00 €

    Specifications:
    More
    Specifications
    Cat. No.
    17-5131-02
    Max. pressure
    5 bar
    pH range
    3 - 12
    Storage
    4 to 30 °C, 20% Ethanol