Order Entry
Germany
ContactUsLinkComponent
VWR® Protease Inhibitor Cocktail, Plant
VWR® Protease Inhibitor Cocktail, Plant
Catalog # M307-1ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Protease Inhibitor Cocktail, Plant
Catalog # M307-1ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    Protease inhibitor cocktail, plant

Specifications

About this item

Contains a mix of protease inhibitors: AEBSF, E-64, Bestatin, Pepstatin, Leupeptin and 1, 10-Phenanthroline. Each vial can be reconstituted in 1 ml dimethyl sulfoxide to form a 100X solution.