Screw caps
Supplier: VITLAB
Jump to:
About this item
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
8 Options Available Below
Product Details & Documents
Made of PP, translucent.
Caps are autoclavable at 121 °C (2 bar) according to DIN EN 285.
Product Family Options
Product Information
- ThreadAvailabilityPrice