Safety ankle boots, lace-up, Alexia
Supplier: Jallatte
Certificates
About this item
Laced shoes made from full grain leather, nylon lining, padded ergonomic collar and dual-density sole.
- Antibacterial nylon lining
- Full piece, anti-static and anatomic integrated insole, perforated in the front part for better foot venting, cushioning effect in the heel area
- TPU/TPU versitale dual-density sole is resistant to slipping and abrasion
- Protective steel toecap (200 J)
- Light, metal-free composite midsole
Colour: Black
Certifications: EN ISO 20345:2007; S3, SRC