Order Entry
Germany
ContactUsLinkComponent
Acriflavine hydrochloride
Acriflavine hydrochloride
Catalog # J60048.14
CAS Number:  
Acriflavine hydrochloride
Catalog # J60048.14
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

About this item

 :
Merck Index:14,123
ADR:9,III
 :578.36 g/mol
UN:3077
 :GHS05
 :GHS07
Storage Temperature:Ambient
CAS Number:8063-24-9
Formula:C₂₇H₂₈Cl₄N₆
MDL Number:MFCD00069039