Order Entry
Germany
Orders LinkContactUsLinkComponent
 

α-Cyclodextrine hydrate

Glass bottle for solids, 100 g
Catalog # UPTIU03449
Supplier:   Uptima
α-Cyclodextrine hydrate
Catalog # UPTIU03449
Supplier:  Uptima
CAS Number:  51211-51-9
α-Cyclodextrine hydrate

Glass bottle for solids, 100 g

CAS Number:  51211-51-9
Price
Inquire For Price
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  α-Cyclodextrin hydrate, α-Cyclodextrin xhydrate
  • CAS Number:51211-51-9
  • Formula:C₃₆H₆₀O₃₀·xH₂O
  • MDL Number:MFCD00150804
  • Molecular Weight:972.86 g/mol (anhy)
  • EINECS:233-007-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Cat. No.:
    UPTIU03449

Specifications