
Choline hydrogen tartrate ≥98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:87-67-2
:
Formula:C₉H₁₉NO₇
MDL Number:MFCD00036332
:253.25 g/mol
Melting Pt:151…153 °C
EINECS:201-763-4
:GHS07
Storage Temperature:Ambient