Order Entry
Germany
ContactUsLinkComponent
Potassium nitrate 99.0-100.5%, granular USP, Multi-Compendial, J.T.Baker®
Oxidizing
Potassium nitrate 99.0-100.5%, granular USP, Multi-Compendial, J.T.Baker®
Catalog # 3193-85
Supplier:  Avantor
CAS Number:  
undefined
Oxidizing
Potassium nitrate 99.0-100.5%, granular USP, Multi-Compendial, J.T.Baker®
Catalog # 3193-85
Supplier:  Avantor
CAS Number:  

Specifications

  • Pack type:
    HDPE pail, Flowmor™
  • Pk:
    20 kg
  • Seq:
    4241

Specifications

Specification Test Results

  • Bulk Food Chemical:
    99.0 - 100.5 %
  • USP - Assay (KNO₃):
    99.0 - 100.5 %
  • USP - Arsenic (As):
    ≤3 ppm
  • USP - Chloride (Cl):
    ≤0.03 %
  • USP - Identification A:
    Passes Test
  • USP - Identification B:
    Passes Test
  • USP - Iron (Fe):
    ≤0.001 %
  • USP - Lead (Pb):
    ≤4 mg/kg
  • USP - Nitrite (NO₂):
    ≤5 ppm
  • USP - Sodium (Na):
    ≤0.1 %
  • USP - Sulfate (SO₄):
    ≤0.1 %
  • JSFA - Arsenic (As):
    ≤2 ppm
  • JSFA - Chloride (Cl):
    ≤0.021 %
  • JSFA - Clarity and Color of Solution:
    Passes Test
  • JSFA - Loss on Drying at 105°C (4 hours):
    ≤1 %
  • EP/BP - Assay (dried basis):
    99.0 - 101.0 %
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Acidity or Alkalinity:
    Passes Test
  • EP/BP - Reducing Substances:
    Passes Test
  • EP/BP - Chloride (Cl):
    ≤20 ppm
  • EP/BP - Sulfate (SO₄):
    ≤150 ppm
  • EP/BP - Ammonium (NH₄):
    ≤100 ppm
  • EP/BP - Calcium (Ca):
    ≤100 ppm
  • EP/BP - Iron (Fe):
    ≤20 ppm
  • EP/BP - Sodium (Na):
    ≤0.10 %
  • EP/BP - Loss on Drying at 105°C:
    ≤0.5 %
  • FCC - Assay (KNO₃) (dried basis):
    99.0 - 100.5 %
  • FCC - Chlorate:
    Passes Test
  • FCC - Identification A:
    Passes Test
  • FCC - Identification B:
    Passes Test
  • FCC - Loss on Drying at 105°C (4 hours):
    ≤1 %
  • No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
    = 1 %

Specification Test Results

About this item

 :
 :101.1 g/mol
Merck Index:13,07733
Melting Pt:334 °C
Density:2.109 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-818-8
UN:1486
ADR:5.1,III
REACH:01-2119488224-35
 :GHS03
CAS Number:7757-79-1
Formula:KNO₃
MDL Number:MFCD00011409