ContactUsLinkComponent
Potassium iodide
Potassium iodide
  RC-152
 :  
Potassium iodide
  RC-152
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024
 :
 :166 g/mol
 :13,07727
 :1330 °C (1013 hPa)
 :686 °C
 :3.13 g/cm³ (20 °C)
 :Ambient
 :231-659-4
 :7681-11-0
 :Kl
 :MFCD00011405