Order Entry
Germany
Orders LinkContactUsLinkComponent
Potassium iodide

Potassium iodide

Glass bottle for solids, 500 g
Catalog # RC-151
Supplier:   G-Biosciences
Potassium iodide
Catalog # RC-151
Supplier:  G-Biosciences
CAS Number:  7681-11-0
Potassium iodide

Glass bottle for solids, 500 g

CAS Number:  7681-11-0
Price
229,00 €
Log in for availability
1 * 500 g
Product Details & Documents
Synonyms:  Dipotassium diiodide, Kali iodide
  • CAS Number:7681-11-0
  • Formula:Kl
  • MDL Number:MFCD00011405
  • Molecular Weight:166 g/mol
  • Merck Index:13,07727
  • Boiling Pt:1330 °C (1013 hPa)
  • Melting Pt:686 °C
  • Density:3.13 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:231-659-4
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.