Glycerine ≥99.5%, Ultrapure
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:56-81-5
:
Formula:HOCH₂CH(OH)CH₂OH
MDL Number:MFCD00004722
:92.09 g/mol
Merck Index:14,4484
Boiling Pt:182 °C (20 mmHg)
Melting Pt:18 °C
Density:1.260
Flash Pt:160 °C (320 °F)
EINECS:200-289-5
:GHS07
Storage Temperature:Ambient