Cytochalasin B
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:14930-96-2
:
Formula:C₂₉H₃₇NO₅
MDL Number:MFCD00077704
:479.62 g/mol
Merck Index:14,2790
Melting Pt:218…221 °C
Storage Temperature:Freezer
EINECS:239-000-2
UN:1544
ADR:6.1,II
:GHS06
:GHS08