Rifampicin
Catalog # MOLE85067949
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:822.95 g/mol
Merck Index:12,08382
Melting Pt:183…188 °C
EINECS:236-312-0
CAS Number:13292-46-1
Formula:C₄₃H₅₈N₄O₁₂
MDL Number:MFCD00151389