Order Entry
Germany
ContactUsLinkComponent
D-(+)-Glucose
D-(+)-Glucose
Catalog # PROORTC-PHR-1000
CAS Number:  
undefined
D-(+)-Glucose
Catalog # PROORTC-PHR-1000
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
Merck Index:14,04459
Melting Pt:146 °C
Density:1.54 g/cm³ (20 °C)
 :180.16 g/mol
Storage Temperature:Ambient
EINECS:200-075-1
CAS Number:50-99-7
 :492-62-6
Formula:C₆H₁₂O₆
MDL Number:MFCD00148912