Order Entry
Germany
ContactUsLinkComponent
Glycine ≥99.5% cell culture reagent
Glycine ≥99.5% cell culture reagent
Catalog # J62407.A1
CAS Number:  
undefined
Glycine ≥99.5% cell culture reagent
Catalog # J62407.A1
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg
  • Seq:
    4024

Specifications

About this item

 :
 :75.07 g/mol
Merck Index:14,4491
Boiling Pt:181 °C (1013 hPa)
Melting Pt:∼ 245 °C (dec.)
Density:1.595
Storage Temperature:Ambient
EINECS:200-272-2
CAS Number:56-40-6
Formula:NH₂CH₂COOH
MDL Number:MFCD00008131