Order Entry
Germany
ContactUsLinkComponent
 
Glycine 99% ACS
 
undefined
Glycine 99% ACS

 

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Sulfated ash:
    ≤0.1 %
  • Titration with HClO4:
    ≥98.5 %
  • Substance darkened by H2SO4:
    Passes test
  • Chloride (Cl):
    ≤0.005 %
  • Sulfate (SO4):
    ≤0.005 %
  • Ammonium (NH4):
    ≤0.005 %
  • Heavy metals (as Pb):
    ≤0.002 %
  • Hydrolyzable substance:
    Passes test