Order Entry
Germany
ContactUsLinkComponent
2,5-Di-tert-butylhydroquinone
2,5-Di-tert-butylhydroquinone
Catalog # EHERC12252500
Supplier:  EHRENSTORFER
CAS Number:  
2,5-Di-tert-butylhydroquinone
Catalog # EHERC12252500
Supplier:  EHRENSTORFER
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    0,25 g
  • Seq:
    3029

Specifications

About this item

 :
 :222.33 g/mol
Boiling Pt:334 °C (1013 hPa)
Melting Pt:212 °C
Density:1.078 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:201-841-8
CAS Number:88-58-4
Formula:C₁₄H₂₂O₂
MDL Number:MFCD00008825