2,2,2,3',5'-Pentafluoroacetophenone
Catalog # FLUO023155-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:210.1000 g/mol
CAS Number:845823-12-3
Formula:C₈H₃F₅O
MDL Number:MFCD01319993