1,3-Dibromo-5-(trifluoromethoxy)benzene, Sigma-Aldrich®
Catalog # SIAL343951-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:319.90 g/mol
:M-Clarity™ quality level = MQ100
CAS Number:207226-31-1
Formula:C₇H₃Br₂F₃O
MDL Number:MFCD00075280