3'-Chloro-2,2,2-trifluoroacetophenone
Catalog # FLUO023154-1G
Supplier: FLUOROCHEM
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:208.5650 g/mol
CAS Number:321-31-3
Formula:C₈H₄ClF₃O
MDL Number:MFCD00461896