Order Entry
Germany
ContactUsLinkComponent
Hydrogen chloride 3 mol/l in cyclopentyl methyl ether, Sigma-Aldrich®
Hydrogen chloride 3 mol/l in cyclopentyl methyl ether, Sigma-Aldrich®
Catalog # SIAL705551-100ML
Supplier:  Merck
CAS Number:  
Hydrogen chloride 3 mol/l in cyclopentyl methyl ether, Sigma-Aldrich®
Catalog # SIAL705551-100ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :36.46 g/mol
Storage Temperature:Ambient
EINECS:231-595-7
UN:2924
ADR:3,II
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324