Order Entry
Germany
ContactUsLinkComponent
4-Fluoro-2-methylphenylboronic acid
4-Fluoro-2-methylphenylboronic acid
Catalog # WAKO328-69071
Supplier:  Wako
CAS Number:  
4-Fluoro-2-methylphenylboronic acid
Catalog # WAKO328-69071
Supplier:  Wako
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :153.95 g/mol
CAS Number:139911-29-8
 :MFCD17214439
Formula:C₇H₈BFO₂
MDL Number:MFCD02093072