Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Silicon nitride
Silicon nitride, Sigma-Aldrich®
Silicon nitride, Sigma-Aldrich®
Silicon nitride, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Silicon nitride, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
12033-89-5
:
Formula:
N₄Si₃
MDL Number:
MFCD00011230
:
140.28 g/mol
:
M-Clarity™ quality level = MQ100
EINECS:
234-796-8