0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Germany
ContactUsLinkComponent
4-Isopropyl-3-methylaniline hydrochloride
4-Isopropyl-3-methylaniline hydrochloride
4-Isopropyl-3-methylaniline hydrochloride
4-Isopropyl-3-methylaniline hydrochloride
Supplier:
Thermo Scientific
Certificates
Expand image
4-Isopropyl-3-methylaniline hydrochloride
Supplier:
Thermo Scientific
About this item
CAS Number:
4534-11-6
:
Formula:
C₁₀H₁₆ClN
MDL Number:
MFCD00156562
:
185.70 g/mol
EINECS:
224-876-0