
4-Formyl-3-(trifluoromethyl)phenylboronic acid 95%
Catalog # H26988.03
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
About this item
:
:217.94 g/mol
Melting Pt:200…202 °C
:GHS07
Storage Temperature:Ambient
CAS Number:1072944-24-1
Formula:C₈H₆BF₃O₃
MDL Number:MFCD09037498