2-(p-Toluenesulphonyl)acetophenone
Catalog # MENANC2279.25
Supplier: MENAI
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
Specifications
About this item
:
:274.340 g/mol
EINECS:250-605-0
CAS Number:31378-03-7
Formula:C₁₅H₁₄O₃S
MDL Number:MFCD00026002