N,N-Bis(trifluoromethanesulphonyl)aniline ≥99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:37595-74-7
:
Formula:C₈H₅F₆NO₄S₂
MDL Number:MFCD00000404
:357.25 g/mol
Melting Pt:100…102 °C
:GHS07
Storage Temperature:Ambient