Order Entry
Germany
ContactUsLinkComponent
N,N-Bis(trifluoromethanesulphonyl)aniline 97%
N,N-Bis(trifluoromethanesulphonyl)aniline 97%
Certificates
undefined
N,N-Bis(trifluoromethanesulphonyl)aniline 97%

Specification Test Results

  • Appearance:
    White to yellow to beige Crystalline powder or crystals and/or chunks
  • Infrared spectrum:
    Conforms
  • Melting point:
    99 to 105 °C
  • HPLC:
    ≥96.0 %

Specification Test Results

Related Information