(S)-(-)-N-Benzyl-α-methylbenzylamine
Catalog # MOLE39744829-5G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:211.31 g/mol
CAS Number:17480-69-2
Formula:C₁₅H₁₇N
MDL Number:MFCD00066325