Divinyl sulphone
Catalog # MSPP140112
Supplier: MARKET SOURCE PART PROCESS
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:118.16 g/mol
EINECS:201-057-6
UN:2927
ADR:6.1,I
CAS Number:77-77-0
Formula:C₄H₆O₂S
MDL Number:MFCD00008623