N,N'-Diallyl-L-tartardiamide, Sigma-Aldrich®
Catalog # SIAL156868-25G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:25 g
- Seq:3308
- Environmentally Preferable:
Specifications
About this item
:
:228.25 g/mol
:M-Clarity™ quality level = MQ100
EINECS:261-277-3
CAS Number:58477-85-3
Formula:C₁₀H₁₆N₂O₄
MDL Number:MFCD00008640