Order Entry
Germany
ContactUsLinkComponent
Hexafluorophosphoric acid 60% in aqueous solution
Hexafluorophosphoric acid 60% in aqueous solution
Catalog # FLUO006568-100G
Supplier:  FLUOROCHEM
CAS Number:  
Hexafluorophosphoric acid 60% in aqueous solution
Catalog # FLUO006568-100G
Supplier:  FLUOROCHEM
CAS Number:  

Specifications

  • Pk:
    100 g
  • Seq:
    3401

Specifications

About this item

 :
 :145.97 g/mol
EINECS:241-006-5
UN:1782
ADR:8,II
CAS Number:16940-81-1
Formula:HF₆P
MDL Number:MFCD00011350