Order Entry
Germany
ContactUsLinkComponent
Carbon disulphide 5 mol/l in THF, Sigma-Aldrich®
Carbon disulphide 5 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL721476-50ML
Supplier:  Merck
CAS Number:  
undefined
Carbon disulphide 5 mol/l in THF, Sigma-Aldrich®
Catalog # SIAL721476-50ML
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    50 mL
  • Seq:
    2370

Specifications

About this item

 :
 :76.14 g/mol
EINECS:200-843-6
CAS Number:75-15-0
Formula:CS₂
MDL Number:MFCD00011321