Order Entry
Germany
ContactUsLinkComponent
Furan-2,5-dicarboxylic acid, Sigma-Aldrich®
Furan-2,5-dicarboxylic acid, Sigma-Aldrich®
Catalog # SIAL722081-5G
Supplier:  Merck
CAS Number:  
Furan-2,5-dicarboxylic acid, Sigma-Aldrich®
Catalog # SIAL722081-5G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :156.09 g/mol
EINECS:221-800-8
CAS Number:3238-40-2
Formula:C₆H₄O₅
MDL Number:MFCD00016582