



Methylmagnesium chloride ∼22% (w/w) 3 mol/l in THF, AcroSeal®
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:676-58-4
:
Formula:CH₃ClMg
MDL Number:MFCD00000468
Boiling Pt:66 °C (760 mmHg)
Density:1.01 g/cm³
Flash Pt:–17 °C
Storage Temperature:Ambient
EINECS:211-629-7
UN:3399
ADR:4.3,I
:74.79 g/mol
:GHS07
:GHS05
:GHS02
:GHS08
Specification Test Results
- Appearance:Dark brown Solution
- Titration with HCl:22 to 24 %