Order Entry
Germany
ContactUsLinkComponent
3-Ethoxy-5-(trifluoromethyl)phenylboronic acid ≥98%
Irritant
3-Ethoxy-5-(trifluoromethyl)phenylboronic acid ≥98%
Catalog # H52989.03
CAS Number:  
Irritant
3-Ethoxy-5-(trifluoromethyl)phenylboronic acid ≥98%
Catalog # H52989.03
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 g

Specifications

About this item

 :
 :233.98 g/mol
Melting Pt:173…176 °C
 :GHS07
Storage Temperature:Ambient
CAS Number:871332-96-6
Formula:C₉H₁₀BF₃O₃
MDL Number:MFCD08056361