Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Germany
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
4-(Trifluoromethylthio)benzyl bromide
4-(Trifluoromethylthio)benzyl bromide
4-(Trifluoromethylthio)benzyl bromide
4-(Trifluoromethylthio)benzyl bromide
Supplier:
CHEMPUR
Certificates
Expand image
4-(Trifluoromethylthio)benzyl bromide
Supplier:
CHEMPUR
About this item
CAS Number:
21101-63-3
:
Formula:
C₈H₆BrF₃S
MDL Number:
MFCD00129189
:
271.10 g/mol
UN:
3261
ADR:
8,II