Order Entry
Germany
Orders LinkContactUsLinkComponent
Hotplate

Hotplate

305×609 mm, 150...371 °C, 3200 W, Hotplate, RC2240Q
Catalog # BARNRC2240Q
Hotplate
Catalog # BARNRC2240Q
Hotplate

305×609 mm, 150...371 °C, 3200 W, Hotplate, RC2240Q

Price
2 090,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

External-controlled hotplates with porcelain-coated stainless-steel top.

  • Isolation of controls allows operation in hazardous environments
  • Large top plate surfaces accommodate up to 40 lb. (18.1 kg) loads
  • Power light indicates when control is operating
  • Large capacity with excellent temperature stability
  • Percentage input controller delivers infinite temperature selection
  • Porcelain-coated, stainless-steel top offers chemical and stain resistance
  • Chemical-resistant cover protects line cord
  • Stainless-steel case offers maximum resistance to corrosion

These hotplates are ideal for a range of applications with acidic or basic aqueous solutions. An external controller operates outside fume hoods and corrosive environment.

Temperature stability is ±5°C (9°F) over the operating temperature range. Embedded heating elements maximize temperature uniformity and evenly transfer heat within ±10°C (18°F) at 150°C (300°F).

Specifications
  • Heating area:
    305×609 mm
  • Heating capacity:
    3200 W
  • Temp.:
    150...371 °C
  • Type:
    Hotplate, RC2240Q