Dishes, evaporating, flat form
Supplier: COWIE
Certificates
About this item
Made of PTFE.
- Very good chemical resistance
- Temperature resistant from -200 to +280 °C
- Exceptionally smooth inner walls to prevent contamination
Thick walled, extremely robust.