Order Entry
Germany
ContactUsLinkComponent
 
Reference substances for GC analysis, Supelco®
 :  Merck
 
undefined
Reference substances for GC analysis, Supelco®
 :  Merck
 

 

  • CAS No.:
    110-42-9
  • M-Clarity quality level:
    MQ100
  • Melting point:
    –18…–10 °C
  • Formula:
    C₁₁H₂₂O₂
  • Description:
    Methyl decanoate