Heat resistant gloves, Aratherma Fit
Ambidextrous knitted gloves made of Kevlar®, with cotton lining and long knitted, elasticated wrist (10 cm). The Kevlar® fibre offers good resistance to cuts and provides protection against thermal hazards (contact heat).
- Knitting technique helps to produce a glove that conforms snugly to the hand and provides good dexterity
- Lining enhances the glove's mechanical performance while absorbing perspiration
- Glove's relative thickness enables it to absorb shocks, making it suitable for the handling of heavy objects
- Long knitted wrist offers forearm protection
- Heat resistant up to 250 °C
Ideal for the handling of hot, sharp parts in a dry atmosphere.
: EN 388:2016 (2X4XD), EN 407 (42XXXX), Cat. II
- Catalog No:
- 112-2891
- 112-2890
- Model:
- Fit
- Fit
- Ambidextrous / hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Reusable
- Reusable
- Liner material:
- Cotton
- Cotton
- Gauge:
- 7
- 7
- Cuff style:
- Knit wrist
- Knit wrist
- Cuff length:
- 10 cm
- 10 cm
- EN norm:
- EN 388:2016 (2X4XD), EN 407 (42XXXX)
- EN 388:2016 (2X4XD), EN 407 (42XXXX)