VWR® Dublin, Safety Spectacles
111-0929
- Type:Anti-UV (2 - 1,2), anti-scratch, anti-fog
- Colour:Transparent
- Seq:01
- Lenses:Clear PC
- Pk:1
Lightweight spectacles with clear polycarbonate lens.
- Anti-scratch/anti-fog coating
- Moulded brow guard
- Soft nose pad
- Soft rubber temple pads
Weight: 25,5 g
EN 166, EN 170; 1 FT
: Compliant with EN 166, EN 170; VWR® EN 166 FT CE (frame); VWR® 2C-1,2 1 FT (lens)