Auftragserfassung
Germany
AuftragsverfolgungKontakt
Capillary columns, Select PAH
Capillary columns, Select PAH
Supplier:   VARIAN
Capillary columns, Select PAH
Supplier:  VARIAN
Certificates
Capillary columns, Select PAH
Capillary columns, Select PAH
2 Options Available Below
Product Details & Documents
Select PAH is the first capillary column that provides a single solution for PAH analysis by separating all the isomers, thereby avoiding false positives and inaccurate results.
  • Provides easy, fast, and accurate quantification of PAHs in environmental and food samples
  • No time wasted on false positives
  • Fast results with no need for further analysis
  • Low bleed reduces maintenance time and enhances sensitivity
The low column bleed of Select PAH reduces the requirement for MS maintenance and enhances sensitivity.
Product Family Options
Product Information
  • Säulenform
    Phase
    Schichtdicke
    Ø int.
    Länge
    Availability
    Price
  • Kapillar
    Select PAH
    0,10 µm
    0,15 mm
    15 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 686,00 €
    686,00 €
    1 * 1 ST
       686,00 €
  • Kapillar
    Select PAH
    0,15 µm
    0,25 mm
    30 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 949,00 €
    949,00 €
    1 * 1 ST
       949,00 €