Orders LinkContactUsLinkComponent
Capillary columns, CP-FFAP CB
Capillary columns, CP-FFAP CB
Supplier:   VARIAN
Capillary columns, CP-FFAP CB
Supplier:  VARIAN
Certificates
Capillary columns, CP-FFAP CB
Capillary columns, CP-FFAP CB
4 Options Available Below
Product Details & Documents
Columns for free fatty acids in dairy products.
  • Ideal for flavors, aromas and free fatty acids C₁ to C₂₆
  • Separates C₂ to C₂₄ acids in one run without derivatisation, saving time
  • Water and solvent resistant
  • Chemically-bonded
Temperature limits: 250/275 °C.
Product Family Options
Product Information
  • Column Format
    Phase
    Film thickness
    Ø int.
    Length
    Availability
    Price
  • 568-0715
    on 7" cage
    on 7" cage
    Capillary
    CP-FFAP CB
    0,25 µm
    0,15 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 815,00 €
    815,00 €
    1 * 1 ST
       815,00 €
  • 568-0698
    on 7" cage
    on 7" cage
    Capillary
    CP-FFAP CB
    0,30 µm
    0,32 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 919,00 €
    919,00 €
    1 * 1 ST
       919,00 €
  • 568-0700
    on 7" cage
    on 7" cage
    Capillary
    CP-FFAP CB
    1,00 µm
    0,53 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 190,00 €
    1 190,00 €
    1 * 1 ST
       1 190,00 €
  • 568-0699
    on 5" cage
    on 5" cage
    Capillary
    CP-FFAP CB
    0,30 µm
    0,32 mm
    25 m
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 150,00 €
    1 150,00 €
    1 * 1 ST
       1 150,00 €