Order Entry
Germany
ContactUsLinkComponent
Capillary columns, CP-Carbowax 400
Capillary columns, CP-Carbowax 400
  VARICP7527
 :  VARIAN
Capillary columns, CP-Carbowax 400
  VARICP7527
 :  VARIAN

 

  • Column Format:
    Capillary
  • Film thickness:
    0,20 µm
  • Ø int.:
    0,32 mm
  • Length:
    50 m
  • Phase:
    CP-Carbowax 400
  • Pk:
    1

 

 

CP-Carbowax 400 has highest resolution for amyl alcohols for accurate quality inspection.

  • High plate number, even at 0 °C, for reliable analysis of the most volatile compounds
  • Specially designed and tested for this application, ensuring ease-of-use
  • Ideal for the analysis of volatiles in alcoholic beverages