Safety ankle boots, lace-up, Jalmars
: Jallatte
Ankle boots made from full grain water repellent leather upper, 3D lining and Softane™/Néotril™ sole (heat resistant up to 300 °C); with padded collar.
- Lining with three dimensional foam structures for excellent ventilation
- Confort-Tech integrated insole: Anatomical, antibacterial, shock absorber heel
- Slip resistant Triftane™ sole helps to prevent fatigue
- Aérfort perforated steel toecap (200 J)
- Puncture resistant stainless steel midsole
Colour: Black
: EN ISO 20345:2007, S3, HRO, SRC.